missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Non-Treated Multidishes
BIOPHARMA

Perform suspension cell cultures with Thermo Scientific™ Nunc™ Non-Treated Multidishes.

Brand:  Thermo Scientific™ 150200

180.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10559532

  • £359.00 / Pack of 75
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Perform suspension cell cultures with Thermo Scientific™ Nunc™ Non-Treated Multidishes.

  • Flat-bottom round wells, with lid
  • Excellent optical properties
  • Raised well rims to lower the risk of cross-contamination
  • Lid with notched corners for good fit and easy orientation
  • Non-pyrogenic
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Multi-Well Plate
12
Sterile
Flat Bottom (F-well)
75
128 mm
2 mL (Well)
2 mL
Non-treated
Lid
1
86 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nunc™ Non-Treated Multidishes > 12 well, Suggested working volume 2.0mL/well; 75/Cs, individually packed

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.