Learn More
Thermo Scientific™ Nunc™ PETG Roller Bottles
Increase cell expansion and product yield while eliminating the need to purchase additional production equipment or increase labor.
£191.00 - £860.00
Specifications
Color Clear
Material PETG
Closure Included Yes
Graduated Yes
Shape Round
View More Specs
Products 8
Product Code Brand Cell Growth Area Bottle Surface Packaging Price Quantity & Availability  
Product Code Brand Cell Growth Area Bottle Surface Packaging Price Quantity & Availability  
10059702
View Documents
Thermo Scientific™
1060-05
1050 cm2 Smooth 5/sleeve, 20/case
£206.00
Pack of 20
In Stock
to see stock.
Add to basket
10712974
View Documents
Thermo Scientific™
1060-20
1050 cm2 Smooth 20/case
£191.00
Pack of 20
In Stock
to see stock.
Add to basket
10263393
View Documents
Thermo Scientific™
1060-85
1050 cm2 Smooth 5/sleeve, 20/case
£246.00
Pack of 20
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
10627584
View Documents
Thermo Scientific™
1760-20
1700 cm2 Pleated 20/case
£341.00
Pack of 20
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
10385073
View Documents
Thermo Scientific™
1860-22
1800 cm2 Smooth 22/case
£447.00
Pack of 22
In Stock
to see stock.
Add to basket
10704144
View Documents
Thermo Scientific™
2160-20
2100 cm2 Pleated 20/case
£422.00
Pack of 20
Estimated Shipment: 11-06-2025
to see stock.
Add to basket
10551844
View Documents
Thermo Scientific™
2160-05
2100 cm2 Pleated 5/sleeve, 20/case
£449.00
Pack of 20
Estimated Shipment: 30-06-2025
to see stock.
Add to basket
10596895
View Documents
Thermo Scientific™
4260-22
4200 cm2 Pleated 22/case
£860.00
Pack of 22
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
Description
Description
Increase cell expansion and product yield while eliminating the need to purchase additional production equipment or increase labor with Thermo Scientific™ Nunc™ PETG Roller Bottles. Virtually unbreakable PETG provides added assurance that valuable contents are safe during laboratory and industrial-scale applications including vaccines, cell culture expansion and production of other biologics.
Key Features:
  • Molded of durable PETG
  • Can be frozen down to -40°C — supports freeze thaw cell release methods, reducing reliance on trypsin
  • Provides solutions for all scales with wide range of surface areas from 1050cm2 to 4200cm2
  • Reduces wrist strain and increases productivity with quick-action ergonomic closure
  • Can be traced by lot number on every bottle
Recommended for:
  • Cell culture expansion
  • Vaccine production
  • Antibody production
Warranty: 90 days
Specifications
Specifications
Clear
Yes
Round
Quick-Action Ergonomic
PETG
Yes
Roller Bottle
Videos
SDS
Documents
Documents
Product Certifications
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Product Title
Thermo Scientific™ Nunc™ PETG Roller Bottles