Learn More
Masterflex™ Nylon Male Luer Lock Plug
Complete your luer fittings systems with these premium connectors. Cole-Parmer™ Nylon Male Luer Lock Plugs are manufactured in compliance with ISO 594-1 standards, inspected to be free from contaminants and are fully traceable.
Brand:  Masterflex™ 45505-56
Product Code. 11790199
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Use these fittings for a variety of critical applications, including syringes or small tubing
  • All parts meet tight dimensional tolerances for increased reliability and process purity
  • Choose from a range of sizes, with a variety of connection options
  • Good chemical resistance. Max temperature 235°C  (455°F)
  • Sterilize by autoclave, ethylene oxide, or gamma irradiation
  • All female ports may be used with either slip or luer lock connections
  • Plugs and caps are also available
Specifications
Specifications
Autoclavable
25/Pk.
235°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Masterflex™ Nylon Male Luer Lock Plug > Male Luer Lock Plug