Learn More
Kern Sohn OIML Class E2 Compact Test Weight
Single test weight constructed of stainless steel.
Brand: Kern Sohn 316-03
Description
- Compact shape
- Polished stainless steel
Specifications
E2 | |
Stainless Steel | |
5 g | |
± 0.05 mg |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.