Learn More
Diba Omnifit™ BenchMark™ Replacement Frits for Chromatography Column Assemblies
Select from a variety of porosities for your media and chemistry
Brand: Diba 006FR-15-25-PE
This item is not returnable.
View return policy
Description
Replacement frits are available in PTFE or PE and in a range of pore sizes, enabling you to select the most appropriate bed supports for your application. Match the column ID to select the correct fritSpecifications
Chromatography Column Assemblies | |
25 μm | |
15 mm | |
Replacement Chromatography Frit |
Polyethylene | |
Universal | |
0.59 in. |