Learn More
Diba Omnifit™ Replacement Glass Tubes for EZ and EZ SolventPlus™ Columns
Mix and match your Omnifit™ Replacement Glass Tubes for EZ and EZ SolventPlus™ Column lengths while using your existing endpieces for ultimate flexibility
Brand:  Diba 006ERG-35-33
Encompass_Preferred
 View more versions of this product
Product Code. 15845304
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
Use a different length tube with your existing endpieces to meet your specific requirements.
Specifications
Specifications
Omnifit™ EZ and EZ SolventPlus™ Columns
Liquid Chromatography Column
Glass
35 mm
330 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Diba Omnifit™ Replacement Glass Tubes for EZ and EZ SolventPlus™ Columns