Learn More
Fisherbrand™ omniPAGE Mini Vertical Electrophoresis System
Suitable for the separation of proteins in polyacrylamide gels.
Brand:  Fisherbrand™ FCVS10DSYS-CU
Product Code. 12347257
  • £560.00 / Each
Estimated Shipment: 09-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • High throughput capability
  • Rapid casting
  • Reversible combs serves as loading indicators which aids pipette-well alignment to prevent sample loading errors
  • Interchangeable modular inserts for slab gels, 2-D electrophoresis and electroblotting
Specifications
Specifications
Vertical Electrophoresis System
omniPAGE Mini, 10 x 10cm Dual, 2 sets of glass plates, Bonded spacers (1mm thick), 2 x 12 sample, combs (1mm thick) including caster, External casting upstand
0.06 to 0.32 gal.
13 x 19 x 15 cm ( 5.12 x 7.48 x 5.90 in.)
1.8 kg
W x H: 8 x 8.5 cm (3.15 x 3.35 in.)
2 to 4
250 to 1200 mL
3.97 lb.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ omniPAGE Mini Vertical Electrophoresis System > omniPAGE Mini Vertical Electrophoresis System