Learn More
Honeywell Safety Products™ OP-TEMA Clear Lens Glasses Product Code.: 11846900
In Stock
to see stock.
Add to basket
Honeywell Safety Products™ OP-TEMA Clear Lens Glasses
Product Code. 11846900
Blue
Click to view available options
Frame Color:
Blue
Product Code. 11846900
Brand: Honeywell Safety Products™ 1000018
In Stock
to see stock.
Add to basket
Add to basket
High-value, affordable eye protection for your busy workplace. Honeywell Safety Products™ OP-TEMA Clear Lens Glasses are designed for both comfort and protection.
  • Added security and convenient protection in your active work environment.
  • Affordably priced protection for maximum value.
  • Lightweight full-view lens.
  • Built-in molded browbar.
  • Soft, adjustable nosepiece.
  • Adjustable temple length and lens angle.
  • Abrasion resistant polycarbonate lens.
  • Adjustable, breakaway Flexicord retainer.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number EC0723, EC Attestation Number EC0723
Lens Coating Impact-resistant, Scratch-resistant, Scratch-resistant, UV-treated
Product Type Safety Glasses
For Use With (Application) Administration, Agriculture, Automotive, Building, Construction, Energy, Electricity, Laboratory, Logistics, Maintenance, Medical, Pharmaceutical, Transportation and Utility industries
Frame Color Blue
No. per Pack 1 Ea.
Weight (Metric) 30 g
Lens Material Polycarbonate
Frame Material Nylon
UV Protection UV400
Product Content Correction
Product Title
Honeywell Safety Products™ OP-TEMA Clear Lens Glasses >