Learn More
Gibco™ Opti-Plex™ Complexation Buffer
Opti-Plex Complexation Buffer is a cGMP-manufactured, chemically defined, animal origin-free (AOF), methionine-free, biotin-free, low-protein transfection complexation buffer based on Opti-MEM I Reduced Serum Medium
Brand: Gibco™ A4096801
37.41 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Designed for complexation of ExpiFectamine transfection reagents for both standard transfection applications and transfections to generate methionine-labeled proteins
Features:
- Contains recombinant insulin, recombinant transferrin, and GlutaMAX Supplement
- Does not contain methionine, biotin, or phenol red
- Part of the ExpiFectamine293 Met(-) Transfection Kit

Specifications
Complexation Buffer | |
Protein Expression | |
Liquid | |
100 mL |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2450039 | Certificate of Analysis | 29/01/2025 | A4096801 |
2450041 | Certificate of Analysis | 29/01/2025 | A4096801 |
2785411 | Certificate of Analysis | 28/01/2025 | A4096801 |
2785413 | Certificate of Analysis | 27/01/2025 | A4096801 |
2898809 | Certificate of Analysis | 27/01/2025 | A4096801 |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.