Learn More
Bel-Art™ SP Scienceware™ Polypropylene Powder Funnel
Polypropylene Powder Funnel - Specially designed for transferring powders with wide stem for clog free filling
Brand: Bel-Art™ SP Scienceware™ H14660-0100
41.33 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Made of polypropylene plastic with 60° angle
- Steam autoclavable at 121°C (250°F)
Specifications
10.0 cm | |
Autoclavable | |
3.25 in. |
248 mL | |
3.94 in. | |
Filling Funnel (Powders) |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Your input is important to us. Please complete this form to provide feedback related to the content on this product.