missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Opti-Link™ Carboxylate-Modified Particles

Control sensitivity, specificity and stability with various acid content available within Thermo Scientific™ Opti-Link™ Carboxylate-Modified Particles.

Brand:  Thermo Scientific™ 83000550100290

 View more versions of this product

Product Code. 12612550

  • £201.00 / 15mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Control sensitivity, specificity and stability with various acid content available within Thermo Scientific™ Opti-Link™ Carboxylate-Modified Particles. Particles contain carboxylic acid groups for covalent coupling and can be used directly from the bottle in a variety of applications.

Parking area combines surface area and acid content to give you the surface acid distribution that is useful for assay optimization. The surface area per gram of a particle varies inversely with the particle diameter. Acid content refers to the titrated milliequivalents per gram (mEq/g) and is weight-based.
  • For most applications, Opti-Link particles can be used directly from the bottle without any pre-cleaning
  • Proprietary anionic surfactant does not interfere with binding of proteins nor cause proteins to desorb from particle surfaces, nor cause proteins to desorb from particle surfaces
  • Various surface acid concentrations available to help optimize reagent development efforts
  • Opti-Link particles have been used in and are equally successful in molecular biology and research applications

Parking Area Properties

  • Low Acid Content—60–100 Parking Area—Hydrophobic surface, more colloidally stable than plain polystyrene
  • Medium Acid Content—35–59 Parking Area—Hydrophobic and hydrophilic areas; good colloidal stability and good covalent coupling
  • High Acid Content—10–34 Parking Area—Hydrophilic surface; excellent colloidal stability; excellent covalent coupling

Recommended for:

Clinical diagnostics, molecular biology, nephelometric, turbidimetric and slide agglutination assays

Note:

When using Opti-Link particles, ask about our Microparticle Reagent Optimization manual, which outlines our reagent development strategy. This manual includes validated binding, coupling and particle reagent optimization protocols.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Proprietary anionic surfactant
10%
Particles that contain carboxylic acid groups for covalent coupling and that can be used in a variety of applications.
Covalent Coupling
Specialty Particles
15 mL
Pasteurized
Packaged in 15mL bottles at 10% solids. The evaluation pack consists of three lots from all available PS or CM particles.
Medium Acid
Includes Certificate of Analysis
Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.
White or Colorless
Polystyrene Sulfate
Package Insert Sheet containing Certicate of Analysis
1.05g/cm³
15 mL
Opti-Link Carboxylate-Modified Polystyrene particles are available in different concentrations and package sizes upon request.
0.2 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number251476 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number83000550100290
Lot Number257716 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number83000550100290
Lot Number273072_STD.STK._113099 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number83000550100290
Lot Number276261_BINDING_SITECEM565_VER_010811_1199 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number83000550100290
Lot Number280085_STD.STK_1199 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog Number83000550100290
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Opti-Link™ Carboxylate-Modified Particles > Dia: 0.2μm; 15mL; Medium acid

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.