Learn More
Macherey-Nagel™ OPTIMA™ 1 GC Capillary Column, 0.5mm O.D.
Designed for separation of components according to boiling points. Macherey-Nagel™ OPTIMA™ 1 GC Capillary Column, 0.5mm O.D. are non-polar phase columns with 100% dimethylpolysiloxane for use in gas chromatography. Available in a range of lengths and film thicknesses.
£252.00 - £980.00
Specifications
Diameter (Metric) 0.5 mm
Diameter (Metric) Inner 0.32 mm
Stationary Phase Optima™ 1
USP Type G1, G2, G38
Type GC Capillary Column
View More Specs
Products 26
Product Code Brand Length (Metric) Film Thickness Price Quantity & Availability  
Product Code Brand Length (Metric) Film Thickness Price Quantity & Availability  
10043011
View Documents
Macherey-Nagel™
726301.10
10 m 0.1 μm
£252.00
Each
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
10796312
View Documents
Macherey-Nagel™
726302.10
10 m 0.25 μm
£252.00
Each
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
10576671
View Documents
Macherey-Nagel™
726323.10
10 m 1 μm
£252.00
Each
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
10154701
View Documents
Macherey-Nagel™
726304.10
10 m 0.5 μm
£252.00
Each
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
10536681
View Documents
Macherey-Nagel™
726302.15
15 m 0.25 μm
£326.00
Each
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
10439741
View Documents
Macherey-Nagel™
726323.15
15 m 1 μm
£326.00
Each
In Stock
to see stock.
Add to basket
10206661
View Documents
Macherey-Nagel™
726301.25
25 m 0.1 μm
£551.00
Each
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
10619001
View Documents
Macherey-Nagel™
726302.25
25 m 0.25 μm
£551.00
Each
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
10114751
View Documents
Macherey-Nagel™
726304.25
25 m 0.5 μm
£551.00
Pack of 25
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
10246471
View Documents
Macherey-Nagel™
726821.25
25 m 0.35 μm
£551.00
Each
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
10556501
View Documents
Macherey-Nagel™
726323.25
25 m 1 μm
£551.00
Each
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
10266701
View Documents
Macherey-Nagel™
726301.30
30 m 0.1 μm
£551.00
Each
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
10194601
View Documents
Macherey-Nagel™
726302.30
30 m 0.25 μm
£551.00
Each
In Stock
to see stock.
Add to basket
10012931
View Documents
Macherey-Nagel™
726304.30
30 m 0.5 μm
£551.00
Pack of 30
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
10451472
View Documents
Macherey-Nagel™
726821.30
30 m 0.35 μm
£551.00
Each
In Stock
to see stock.
Add to basket
10327971
View Documents
Macherey-Nagel™
726301.50
50 m 0.1 μm
£924.00
Pack of 50
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
10206421
View Documents
Macherey-Nagel™
726323.30
30 m 1 μm
£551.00
Each
In Stock
to see stock.
Add to basket
10503792
View Documents
Macherey-Nagel™
726302.50
50 m 0.25 μm
£924.00
Each
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
10505381
View Documents
Macherey-Nagel™
726821.50
50 m 0.35 μm
£924.00
Each
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
10566681
View Documents
Macherey-Nagel™
726304.50
50 m 0.5 μm
£924.00
Pack of 50
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
10640532
View Documents
Macherey-Nagel™
726323.50
50 m 1 μm
£924.00
Pack of 50
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
10556331
View Documents
Macherey-Nagel™
726301.60
60 m 0.1 μm
£980.00
Each
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
10629191
View Documents
Macherey-Nagel™
726302.60
60 m 0.25 μm
£980.00
Each
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
10308261
View Documents
Macherey-Nagel™
726304.60
60 m 0.5 μm
£980.00
Each
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
10351292
View Documents
Macherey-Nagel™
726323.60
60 m 1 μm
£980.00
Each
In Stock
to see stock.
Add to basket
10206571
View Documents
Macherey-Nagel™
726821.60
60 m 0.35 μm
£980.00
Each
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
Description
Description
Nonpolar phase composed of 100% dimethylpolysiloxane for use in gas chromatography.
  • Similar phases: OV-1, DB-1, SE-30, HP-1, SPB-1, CP-Sil 5 CB, Rtx-1, 007-1, BP1, MDN-1, AT-1, ZB-1, OV-101
  • Each column is individually tested and supplied with test certificate
  • Maximum temperatures: 340°C for isothermal operation, 360°C for short isotherms in a temperature program
Specifications
Specifications
0.5 mm
Optima™ 1
GC Capillary Column
1 Each
0.32 mm
G1, G2, G38
340°C to 360°C
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Macherey-Nagel™ OPTIMA™ 1 GC Capillary Column, 0.5mm O.D.