Learn More
Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents
Minimize the risks of improper measurement and contamination with these chemistry reagents, which are contained in self-filling vials.
Brand:  Thermo Scientific™ AC4016
 View more versions of this product
Product Code. 11782198
  • £71.79 / 30 tests
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Minimize the risks of improper measurement and contamination with Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents, which are contained in self-filling vials. Use these reagents with the Orion™ AQUAfast AQ4000 colorimeter to simplify sample preparation and tests. The auto-test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.
  • No. of test: 30
  • Consistency and reproducibility improved with AQUAfast ampoules and an AQ4000 meter
  • Thermo Scientific™ Auto-ID™ eliminates measurement errors
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sulfide
Orion AQUAfast AQ4000 Colorimeter
30 Auto-ID Ampoules
Colorimetric water and wastewater testing equipment
0.2 to 3 mg/L S
30 Ampoules
Ampoule
AQUAfast sulfide (Acid Soluble) Auto-ID ampoules, 30 pack
Methylene Blue
AQUAfast Zero Vial Auto-Test Kit (Cat. No. AQ4ZER)
Wastewater: Standard Methods 4500-S2- D
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents >
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.