Learn More
Thermo Scientific™ Owl™ Leveling Platform
Accommodate most applications with this three-point leveling platform which ensures a flat casting and running surface.
Brand: Thermo Scientific™ BBL-1
Code : NEW
Additional Details : Weight : 2.00000kg
Description
Accommodate most applications with the Thermo Scientific™ Owl™ Leveling Platform, a three-point leveling platform that ensures a flat casting and running surface.
- Size: 36 x 46cm
- Includes bubble level
Specifications
Owl System | |
Owl | |
Bubble Level |