Learn More
Thermo Scientific™ Orion™ Dual Star™ pH, ISE, mV, ORP and Temperature Dual Channel Benchtop Meter

Accurately monitor pH, ion concentration, mV, ORP and temperature on two channels with our pH/ISE benchtop meter for advanced lab analysis.
Brand: Thermo Scientific™ 2115102
Description
Double your measurement capabilities with the Thermo Scientific™ Orion™ Dual Star™ pH and ISE Benchtop Meter. Record accurate and reliable pH, ion concentration, mV, ORP and temperature measurements on two available meter channels, ideal for high throughput pH and ion analysis in the lab. Perform up to 6 point pH and ISE calibrations, log up to 1000 data point sets with time/date stamp and easily transfer to a printer or computer. Use the two channels to measure a pH sample and ISE sample, two pH samples or two ISE samples simultaneously or view each channel separately.
The Orion Dual Star pH/ISE dual channel benchtop meter accelerates sample measurement throughput by showing the results of two channels simultaneously on its large, backlit display. Measure pH, ion concentration (ISE), mV, relative mV or ORP with temperature on two channels using the meter’s two BNC, reference pin-tip and ATC temperature inputs. Collect critical measurements using an assortment of read modes and onscreen ready indicator that alerts when readings are stable. The meter’s Smart Stability and Smart Averaging functions automatically compensate for measurement conditions, so the meter displays more accurate results faster.
- View large, individual measurements on a full screen display or quickly toggle to a split screen display to view two measurements at the same time
- Easy-to-follow onscreen text prompts for calibration and setup menus available in a multi-language user interface with English, Spanish, French, German or Italian options
- EZ Startup menu facilitates quick and easy first-time meter setup
- Numeric keypad and menu-specific function keys allow for quick and efficient meter setup and data entry
- Selectable read modes include Auto-Read, On-Ready, At Timed Intervals, Continuous, Value Change in Measurement and Timed for a variety of measurement collection methods for different analytical methods
- Perform up to a 6 point pH calibration with automatic recognition for USA/NIST and EURO/DIN buffers with option to manually input custom buffer values
- Perform up to a 6 point ion concentration (ISE) calibration with non-linear auto-blank and low-concentration stability options or use incremental methods for single/double known addition and single/double known subtraction
- Non-volatile meter memory holds up to 1000 data points with time and date stamp
- Easily transfer data via USB and RS232 to a printer or computer with free Orion Dual Star Com communication software
- Stir samples with ease using the meter-controlled Orion Star stirrer probe
- Meter-attachable electrode stand assists with moving and maintaining electrodes
- IP54-rated housing handles splashes and is wall-mountable to save space
Warranty: 3-year meter replacement warranty
Services
3-year meter warranty

Specifications
Universal 50 to 60 Hz, 100 to 240 VAC power adapter (Cat. No. 1010003) | |
USB, RS232 | |
±0.002 | |
Segmented (point-to-point) slope, non-linear selectable auto-blank, low concentration range stabilit | |
Yes, adjustable | |
1 point relative mV (RmV) mode, 1 point ORP mode | |
CE, TUV 3-1, FCC Class A | |
1000 with date and time stamp | |
2 to 6 | |
pH, mV, Relative mV, ORP, Ion Concentration (ISE) with Temperature | |
±1999.9 mV | |
0.0001 minimum, 1 to 3 significant digits (user selectable) | |
-5.0°C to +105.0°C | |
Dual Star pH/ISE dual channel meter, 8157BNUMD ROSS Ultra Triode epoxy-body pH/ATC electrode, universal power adapter, literature, computer interface cable and meter test certificate | |
7.000 pH (adjustable), 1.000 ISE (adjustable) | |
English, Spanish, French, Italian and German | |
Non-volatile memory preserves data log, calibration log and meter settings | |
Orion Dual Star pH/ISE/mV/RmV/ORP/Temperature Dual Channel Benchtop Meter | |
pH electrode, ORP electrode, ion selective electrode (ISE), reference half-cell electrode, ATC temperature probe, stirrer probe (includes 8157BNUMD ROSS Ultra Triode pH/ATC electrode) | |
±1999.9 mV | |
0.1 mV | |
Smart Stability and Smart Averaging | |
5°C to 45°C (ambient) | |
Manual or automatic with ATC temperature probe | |
24 x 17 x 9 cm (9.5 x 6.7 x 3.5 in.) | |
0.9 kg | |
90 to 260 VAC |
±0.2 mV or ±0.05% of reading whichever is greater | |
±0.2 mV or ±0.05% of reading whichever is greater | |
±0.1°C | |
High/low limit alarm | |
pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ | |
1 to 6 | |
Transfer single, range or all data points to printer or computer | |
Orion Dual Star pH/ISE dual channel benchtop meter with ROSS Ultra Triode epoxy-body pH/ATC electrode | |
2 | |
0.0001 to 19,900 in ppm, M, mg/L, % or unit-less | |
-2.000 to 19.999 | |
0.1, 0.01, 0.001 | |
Graphic LCD with Backlight | |
Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe) | |
Numeric with menu-specific function keys and dual purpose scroll/shortcut keys | |
Automatic data logging with Auto-Read, On Ready, At Timed Intervals, Value Change In Measurement and | |
10 per channel with password protection | |
Universal AC adapter (included) | |
±1999.9 mV | |
5 to 85 %, non-condensing | |
0.1°C, 0.1°F | |
41°F to 113°F (ambient) | |
1 point temperature offset calibration | |
3 Year Meter Warranty | |
2 lb. | |
90 to 260 VAC 50/60 Hz |
Videos
-
Thermo Scientific Orion meters Taking water analysis to the next level
-
Thermo Scientific Orion meters Taking water analysis to the next level
Special Offers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
Your input is important to us. Please complete this form to provide feedback related to the content on this product.