missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Total Alkalinity Test Kit

Save time by eliminating additional equipment set-up, titrations and calculations with this total alkalinity test kit that provides two-step direct measurement.

Brand:  Thermo Scientific™ 700011-WA


 View more versions of this product

Product Code. 11787767

  • £203.42 / Pack of 4

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Save time by eliminating additional equipment set-up, titrations and calculations. The Thermo Scientific™ Orion™ Total Alkalinity Test Kit provides two-step direct measurement of total alkalinity and pH using a pH electrode.

  • Direct reading of total alkalinity from 0 to 200ppm as CaCO3, from the enclosed pH conversion chart
  • No interference from color or turbidity

Includes:

  • One 1 pint (475mL) bottle alkalinity reagent (700011)
  • One 1 pint (475mL) bottle alkalinity standard/control (700012)
  • Total alkalinity conversion chart
  • Application procedure No. 517, Total Alkalinity in Natural Waters

Note:

The following part number has been changed due to a recent system upgrade.

Old Part Number: 700011

New Part Number: 700011-WA

Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Safety Data Sheet (MSDS) files.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Alkalinitiy Reagent Refill for Total Alkalinity Test Kit
Liquid
pH Electrode
4 x 475mL
Water, Succinic Acid, Chloroacetic Acid, Sodium Hydroxide, Hyamine 1622, Potassium Chloride
3 to 5
measurement of total alkalinity
Bottle
Total Alkalinity Test Kit
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Total Alkalinity Reagent Solution

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.

Supplemental information
  • MIXTURE LIST-Contains : potassium chloride,succinic acid, chloroacetic acid, sodiumhydroxide

SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberCO1 Certificate TypeCertificate of Analysis Date06/01/2025 Catalog Number700011-WA
Lot NumberCX1 Certificate TypeCertificate of Analysis Date27/03/2024 Catalog Number700011-WA
Lot NumberCY1 Certificate TypeCertificate of Analysis Date06/02/2024 Catalog Number700011-WA
Lot NumberBV1 Certificate TypeCertificate of Analysis Date26/05/2023 Catalog Number700011-WA
Lot NumberAQ1 Certificate TypeCertificate of Analysis Date28/10/2022 Catalog Number700011-WA
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Orion™ Total Alkalinity Test Kit > Alkalinitiy Reagent Refill for Total Alkalinity Test Kit, 4 x 475mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.