missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osmium(III) chloride trihydrate, Premion™, 99.99% (metals basis), Os 52-56%, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.25 g
1 g
5 g
Description
Osmium(III) chloride trihydrate is used as a reagent for preparation of dichlorodihydridoosmium complexes. It is also useful as starting materials and an efficient alkane oxidation catalyst in the presence of pyridine and hydrogen oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | (decomposition) |
Color | Black-Red |
Physical Form | Crystalline |
Quantity | 0.25 g |
Linear Formula | OsCl3·3H2O |
UN Number | UN3260 |
Solubility Information | Soluble in water. |
Formula Weight | 350.60 (296.56 Anhydrous) |
Percent Purity | 99.99% |
Grade | Premion™ |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'