Learn More
Osmium(IV) oxide, Os 83% min, Thermo Scientific Chemicals
Description
The osmium oxide/hexacyanoruthenate films modified electrodes plays an important role for the electrocatalytic reaction of catecholamines and sulfur oxoanions. Used as catalyst. Metallic osmium or an osmium oxide-ruthenium cyanide film modified graphite electrodes plays an essential role for the determination of biogenic amines from electrocatalytic responses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Black |
Physical Form | Solid |
Quantity | 0.25 g |
Solubility Information | Slightly soluble in cold water. |
Formula Weight | 222.2 |
Appearance | Black powder |
Density | 11.4 |
Chemical Name or Material | Osmium(IV) oxide |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
W21J005 | Certificate of Analysis | 24/10/2023 | 039497.06,039497.03,039497.01 |
X11I010 | Certificate of Analysis | 18/10/2022 | 039497.06,039497.03,039497.01 |
Y15H034 | Certificate of Analysis | 12/04/2022 | 039497.06,039497.03,039497.01 |
B28Z013 | Certificate of Analysis | 08/12/2021 | 039497.06,039497.03,039497.01 |
H19U012 | Certificate of Analysis | 08/12/2021 | 039497.06,039497.03,039497.01 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.