missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osmium tetroxide, 2.5 wt.% sol. in tert-Butanol, stab., AcroSeal™, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent

Specifications
Specifications
CAS | 20816-12-0, 75-65-0 |
Quantity | 100 mL |
Molecular Formula | O4Os |
Linear Formula | OsO4 |
MDL Number | MFCD00011150 |
Flash Point | 4°C |
Merck Index | 15, 6990 |
Synonym | osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4 |
InChI Key | VUVGYHUDAICLFK-UHFFFAOYSA-N |
SMILES | O=[Os](=O)(=O)=O |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0401379 | Certificate of Analysis | 14/12/2022 | 429081000 |
A0406213 | Certificate of Analysis | 13/12/2022 | 429081000 |
A0445939 | Certificate of Analysis | 13/09/2022 | 429081000 |
A0438112 | Certificate of Analysis | 27/07/2022 | 429081000 |
A0431901 | Certificate of Analysis | 07/07/2021 | 429081000 |
Safety and Handling




Product Identifier
- Osmium tetroxide, 2.5 wt% solution in t-butanol, stabilized
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Germ cell mutagenicity Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H314-Causes severe skin burns and eye damage.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H341-Suspected of causing genetic defects.
- H301-Toxic if swallowed.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Osmium tetroxide, 2.5 wt.% sol. in tert-Butanol, stab., AcroSeal™, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction