missing translation for 'onlineSavingsMsg'
Learn More
Osmium tetroxide, 99.9+%, (trace metal basis), Thermo Scientific Chemicals Product Code.: 10531641

Osmium tetroxide, 99.9+%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 10531641
250 mg
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 10531641

Brand: Thermo Scientific Acros 191182500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis, Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20816-12-0
Molecular Formula O4Os
Molecular Weight (g/mol) 254.23
MDL Number MFCD00011150
InChI Key VUVGYHUDAICLFK-UHFFFAOYSA-N
Synonym osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4Show MoreShow Less
PubChem CID 30318
IUPAC Name tetraoxoosmium
SMILES O=[Os](=O)(=O)=O

Specifications

Boiling Point 130.0°C
Melting Point 40.0°C to 42.0°C
Color Colorless to Yellow
Physical Form Crystals
Quantity 250 mg
Assay Percent Range 99.9+%
Linear Formula OsO4
Fieser 01,759; 02,301; 04,361; 05,141; 06,424; 10,290; 12,358; 13,186; 14,235; 15,240; 16,249; 17,236
Merck Index 15, 6990
Solubility Information Passes test
Formula Weight 254.2
Percent Purity ≥99.9%
Grade Trace Metal Basis
Packaging Ampoule
Chemical Name or Material Osmium tetroxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467183 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number191185000,191182500,191181000,191180010,191180050
Lot NumberA0462168 Certificate TypeCertificate of Analysis Date20/05/2024 Catalog Number191185000,191182500,191181000,191180010,191180050
Lot NumberA0457102 Certificate TypeCertificate of Analysis Date20/11/2023 Catalog Number191185000,191182500,191181000,191180010,191180050
Lot NumberA0457180 Certificate TypeCertificate of Analysis Date15/11/2023 Catalog Number191185000,191182500,191181000,191180010,191180050
Lot Number4008-232202 Certificate TypeCertificate of Analysis Date29/09/2023 Catalog Number191185000,191182500,191181000,191180010,191180050
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Osmium(VIII)-tetroxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P330-Rinse mouth.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Osmium tetroxide, 99.9+%, (trace metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.