missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osmium Tetroxide, ACS Reagent, ≥98%, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Boiling Point | 130°C |
Melting Point | 39.5°C to 41°C |
Physical Form | Solid |
Quantity | 1 g |
Linear Formula | OsO4 |
UN Number | UN2471 |
Solubility Information | Slightly soluble in water |
Formula Weight | 254.23g/mol |
Percent Purity | ≥98% |
Odor | Unpleasant |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Osmium Tetroxide, ACS Reagent, ≥98%, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction