missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Product Code | Brand | Product Type | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||||
11927535
|
Thermo Scientific™
CM-10 |
Cellophane, Precut | 14 cm | 14 cm | N/A | N/A | N/A | |||||
11708174
|
Thermo Scientific™
CM-20 |
Cellophane, Precut | 24 cm | 24 cm |
£53.25
Pack of 50 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
||||||
11937535
|
Thermo Scientific™
GDF-1 |
Gel Drying Frame | 24 cm | 24 cm | N/A | N/A | N/A | |||||
11917535
|
Thermo Scientific™
GDF-3 |
Gel Drying Frame | 14 cm | 14 cm |
£31.25
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
Description
Combine the frames and pre-cut cellophane that come in Thermo Scientific™ Owl™ Gel-Drying Supplies with Owl™ Gel-Drying Kits.
- Frames and pre-cut cellophane available in large (24 × 24cm) or small (14 × 14cm) format
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Thermo Scientific™ Owl™ Gel-Drying Supplies