Learn More
WTW™ OxiTop™-i Respirometric BOD Measuring System
The WTW™ OxiTop™-i Respirometric BOD Measuring System is designed for self-check BOD measurement with undiluted samples (according to DIN EN 1899 -2) at measuring ranges up to 4000 mg/L.
Brand:  WTW™ 209700
430.35 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
 View more versions of this product
Product Code. 16458763
  • £474.00 / Each
Explore more special offers
Description
Description
  • OxiTop™ is a respirometric vessel-top measuring system for BOD self-check measurement with undiluted samples (DIN EN 1899 -2)
  • Measuring ranges up to 4000 mg/L BOD possible
  • No need to convert data - direct input of sample volume
  • Direct display of the BOD value on the measuring head screen
  • Menu-operated, with backlit graphic LCD, control LED and operating buttons for direct BOD readings, volume pre-selection, measurement duration between 1 and 7 days selectable, display of measuring curve, display of current readings
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OxiTop™-IDS 1
Backlit Graphic LCD
Measuring ranges up to 4000 mg/L BOD possible
One OxiTop™-IDS measuring head