missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brilliance Bacillus Cereus Selective Supplement

Brilliance is a chromogenic medium for the isolation and differentiation of Bacillus cereus in food samples.

Brand:  Thermo Scientific™ SR0230E

77.56 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 13265409

  • £81.39 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Brilliance is a chromogenic medium for the isolation and differentiation of Bacillus cereus in food samples.

Add to Brilliance Bacillus Cereus Agar Base, Part No. CM1036A and CM1036B. Each vial supplements 500mL of medium.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Bacillus cereus
Freeze-dried
10/Pk.
Brilliance Bacillus Cereus Selective Supplement
Selective Supplement
Each vial for 500mL medium
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6244624 Certificate TypeCertificate of Analysis Date07/04/2025 Catalog NumberSR0230E
Lot Number6180774 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog NumberSR0230E
Lot Number6150840 Certificate TypeCertificate of Analysis Date16/08/2024 Catalog NumberSR0230E
Lot Number6116059 Certificate TypeCertificate of Analysis Date18/06/2024 Catalog NumberSR0230E
Lot Number3770249 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog NumberSR0230E
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Brilliance Bacillus Cereus Selective Supplement > 10/Pk.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.