missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Bacillus Cereus Selective Supplement

Oxoid Bacillus Cereus Selective Supplement (Polymyxin B Supplement) is supplied 50,000 IU/Vial for inhibition of Gram-negative bacteria.

Brand:  Thermo Scientific™ SR0099E

58.71 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 11404179

  • £114.00 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Oxoid Bacillus Cereus Selective Supplement (Polymyxin B Supplement) is supplied 50,000 IU/Vial for inhibition of Gram-negative bacteria.

  • May be added to Bacillus cereus Selective Agar Base, Part No. CM0617B and MYP Agar Base, Part No. CM0929B
  • Each vial supplements 500mL of medium
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Bacillus cereus
Freeze-dried
10 x 50,000 IU
Bacillus Cereus Selective Supplement
Selective Supplement
Each vial for 500mL medium
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6243914 Certificate TypeCertificate of Analysis Date08/04/2025 Catalog NumberSR0099E
Lot Number6220816 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberSR0099E
Lot Number6230019 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberSR0099E
Lot Number6202689 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog NumberSR0099E
Lot Number6169786 Certificate TypeCertificate of Analysis Date17/12/2024 Catalog NumberSR0099E
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Oxoid™ Bacillus Cereus Selective Supplement > 10 vials; for 500mL medium

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.