missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MYP Agar (Dehydrated)

Enumerate Bacillus cereus in food samples with this selective and differential medium.

Brand:  Thermo Scientific™ CM0929B

94.71 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10769413

  • £194.00 / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Enumerate Bacillus cereus in food samples with selective and differential medium Thermo Scientific™ Oxoid™ MYP Agar (Dehydrated). The diagnostic features of the medium rely upon the failure of Bacillus cereus to utilize mannitol and the ability of most strains to produce phospholipase C, resulting in an opaque halo around the colonies.

Bacillus cereus has been recognized as a causative agent of food poisoning since the 1950s. Many early isolation techniques used blood agar, relying on hemolysis and colony morphology, followed by a range of tests for confirmation. MYP Agar has proven to be very effective for detecting B. cereus even with very low numbers.

  • Selective: due to the presence of Polymyxin B the medium inhibits most Gram-negative bacteria.
  • Easy-to-read: easy to differentiate Bacillus cereus (opaque halo zone of precipitation around the colonies) from other Gram-positive organisms (no zones of inhibition).

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Industrial Reference: AFNOR, AOAC, BSI, FDA, ISO, NMKL.
Powder
500 g
MYP Agar
Agar
For 11.6L medium
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6224051 Certificate TypeCertificate of Analysis Date28/02/2025 Catalog NumberCM0929B
Lot Number6196814 Certificate TypeCertificate of Analysis Date05/12/2024 Catalog NumberCM0929B
Lot Number6168760 Certificate TypeCertificate of Analysis Date18/10/2024 Catalog NumberCM0929B
Lot Number6157390 Certificate TypeCertificate of Analysis Date29/08/2024 Catalog NumberCM0929B
Lot Number6137421 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog NumberCM0929B
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ MYP Agar (Dehydrated) > Mannitol Yolk Polymyxin B (MYP) Agar; 500g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Laboratory Use Only