Learn More
SymLine™ Signal Boxes
Monitor two filling levels at once. Ideal for integration into lab furniture with SymLine™ Built-in Signal Box.
Brand:  SymLine™ 108124
 View more versions of this product
Product Code. 15965801
Description
Description
  • Clever way to change disposal container without interrupting the inflow
  • LEDs and acoustic buzzer warn you before filling level of collection containers reaches critical height
  • When container is changed, alert can be muted via new touchpad
Specifications
Specifications
Signalbox T5
7.1 in.
UK-style
24 V
Monitoring Filling Levels
T5 with 5 Sensor Inputs and 6 Potential-free Outputs
SCAT™ Sensors and Safety Waste Caps
55 mm
180 x 105 x 55 mm
2 A
105 mm
180 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SymLine™ Signal Boxes > Signal Boxes; Includes: T5 with 5 Sensor Inputs and 6 Potential-free Outputs; Length (Metric): 55 mm; Type: Signalbox T5; Plug Type: UK-style; Voltage: 24 V