Learn More
Thermo Scientific™ Oxoid™ IMAGEN™ Chlamydia Positive Control Slide
Oxoid IMAGEN Chlamydia Positive Control Slide contains acetone-fixed human epithelial cells infected with Chlamydia.
Brand: Thermo Scientific™ S610930-2
81.57 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The IMAGEN™ Chlamydia Control Slides are for use as a control for the IMAGEN™ Chlamydia test (Code No. K610111-2).
Sealed in foil pouch filled with nitrogen.Specifications
IMAGEN™ Chlamydia Positive Control Slide | |
Chlamydia | |
Chlamydia Control Slide | |
1 Slide/Pk. |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.