Learn More
Thermo Scientific™ Mueller Hinton Broth (Dehydrated)
Perform effortless Antimicrobial Susceptibility Testing (AST) using this medium which may be used in internationally recognized standard procedures
Brand: Thermo Scientific™ CM0405B
137.27 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Perform effortless Antimicrobial Susceptibility Testing (AST) using Thermo Scientific™ Oxoid™ Mueller Hinton Broth (Dehydrated), a medium that can be used in internationally recognized standard procedures.
The major use of Mueller Hinton Broth is for Antimicrobial Susceptibility Testing (AST). It has been produced in parallel with Mueller-Hinton Agar CM0337. Where studies on antibiotic susceptibilities are being undertaken both in broth and agar, it will be found to be of particular value to have media of identical nutrient formulation. Mueller-Hinton Broth is recommended for broth dilution Minimum Inhibitory Concentration (MIC) studies1.
Mueller-Hinton Broth will require supplementation with the divalent cations Mg++ and Ca++ after sterilization2. The CLSI recommend the following cation levels Ca++, 20-25mg/L; Mg++, 10-12.5mg/L.
Lysed horse blood or thymidine phosphorylase may be added to the broth to improve the MIC endpoints of sulphonamides and trimethoprim3.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

Specifications
Conforms to CLSI standards. | |
Powder | |
500 g |
Mueller Hinton Broth | |
Dehydrated Culture Media | |
For 23.8L medium |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Laboratory Use Only