Learn More
Thermo Scientific™ Oxoid Novobiocin Selective Supplement, Liquid, 10 mg
Selective supplement for variety of culture media.
Brand: Thermo Scientific™ SR0249A
146.30 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
Oxoid Novobiocin Selective Supplement (10mg/vial) can be added to a range of culture media such as:
BPW (ISO formulation) Part number CM1211 as part of the Salmonella Precis method EC Broth, Part No. CM0990B; Muller-Kauffmann Tetrathionate-Novobiocin Broth Base, Part No. CM0343B; Brilliant Green Agar, Part No. CM0263B; MRSV, Part No. CM1112B; Modified Tryptone Soya Broth, Part No. CM0989B; Lysine Iron Agar, Modified, Part No. CM0886B
Each vial contains 40 mL of liquid novobiocin that has a concentration of 10 mg.

Specifications
Liquid | |
Microbiology Media | |
10 Vials |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6235715 | Certificate of Analysis | 07/03/2025 | SR0249A |
6229594 | Certificate of Analysis | 19/02/2025 | SR0249A |
6229593 | Certificate of Analysis | 19/02/2025 | SR0249A |
6229592 | Certificate of Analysis | 19/02/2025 | SR0249A |
6229591 | Certificate of Analysis | 19/02/2025 | SR0249A |
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.