Learn More
Thermo Scientific™ Oxoid™ Salmonella Selective Supplement
Oxoid Salmonella Selective Supplement is used for the presumptive identification of Salmonella species from clinical and food samples.
Brand: Thermo Scientific™ SR0194E
76.79 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Oxoid Salmonella Selective Supplement is used for the presumptive identification of Salmonella species from clinical and food samples.
- Add to Chromogenic Salmonella Agar Base, Part No. CM1007B or Brilliance™ Salmonella Agar Base, Part No. CM1092B
- Each vial supplements 500mL of medium

Specifications
Salmonella species | |
Freeze-dried | |
10/Pk. |
Salmonella Selective Supplement | |
Selective Supplement | |
Each vial for 500mL medium |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- SALMONELLA Selective Supplement
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : cefsulodin, novobiocin
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6230025 | Certificate of Analysis | 26/02/2025 | SR0194E |
6208066 | Certificate of Analysis | 20/01/2025 | SR0194E |
6199412 | Certificate of Analysis | 04/12/2024 | SR0194E |
6189404 | Certificate of Analysis | 11/11/2024 | SR0194E |
6169124 | Certificate of Analysis | 20/09/2024 | SR0194E |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.