missing translation for 'onlineSavingsMsg'
Learn More
p-Sexiphenyl, 95%, Thermo Scientific Chemicals Product Code.: 15443497

p-Sexiphenyl, 95%, Thermo Scientific Chemicals

Product Code. 15443497
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15443497

Brand: Thermo Scientific Alfa Aesar H26586.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4499-83-6
Molecular Formula C36H26
Molecular Weight (g/mol) 458.604
MDL Number MFCD00039559
InChI Key ZEMDSNVUUOCIED-UHFFFAOYSA-N
Synonym p-sexiphenyl, p-hexaphenyl, p-quaterphenyl, 4,4'-diphenyl, 1-phenyl-4-4-4-4-phenylphenyl phenyl phenyl benzene, p-sexiphenylene, acmc-209k0d, phthalocyanine lead ii purified by sublimation, 1,1':4',1:4,1'-quaterphenyl, 4,4'-diphenyl, 4,4'-bis 1,1'-biphenyl-4-yl-1,1'-biphenylShow MoreShow Less
PubChem CID 78254
IUPAC Name 1-phenyl-4-[4-[4-(4-phenylphenyl)phenyl]phenyl]benzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)C4=CC=C(C=C4)C5=CC=C(C=C5)C6=CC=CC=C6

Specifications

Melting Point >400°C
Quantity 250 mg
Formula Weight 458.59
Percent Purity 95%
Chemical Name or Material p-Sexiphenyl

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10145544 Certificate TypeCertificate of Analysis Date26/07/2023 Catalog NumberH26586.03,H26586.MD
Lot Number10145544 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26586.03,H26586.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
p-Sexiphenyl, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.