missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals

£20.20 - £430.14

Chemical Identifiers

CAS 98-59-9
Molecular Formula C7H7ClO2S
Molecular Weight (g/mol) 190.64
MDL Number MFCD00007450
InChI Key YYROPELSRYBVMQ-UHFFFAOYSA-N
Synonym tosyl chloride, p-toluenesulfonyl chloride, p-tosyl chloride, 4-toluenesulfonyl chloride, p-toluenesulfochloride, 4-methylbenzene-1-sulfonyl chloride, p-tolylsulfonyl chloride, p-toluenesulphonyl chloride, tosylchloride, p-toluenesulfonic acid chloride
PubChem CID 7397
IUPAC Name 4-methylbenzenesulfonyl chloride
SMILES CC1=CC=C(C=C1)S(Cl)(=O)=O
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10736161
View Documents
Thermo Scientific Acros
139030250
25 g Glass bottle
£20.50
25g
Please to purchase this item. Need a web account? Register with us today!
10443883
View Documents
Thermo Scientific Acros
139031000
100 g Glass bottle
£20.20
100g
Please to purchase this item. Need a web account? Register with us today!
10437720
View Documents Promotion Details
Thermo Scientific Acros
139035000
500 g Glass bottle
£49.45
500g
Please to purchase this item. Need a web account? Register with us today!
10775981
View Documents
Thermo Scientific Acros
139030020
2 kg Glass bottle
£128.64
2kg
Please to purchase this item. Need a web account? Register with us today!
11944581
View Documents
Thermo Scientific Acros
139030100
10 kg Plastic drum
£430.14
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

98-59-9
190.64
YYROPELSRYBVMQ-UHFFFAOYSA-N
7397
CC1=CC=C(C=C1)S(Cl)(=O)=O
C7H7ClO2S
MFCD00007450
tosyl chloride, p-toluenesulfonyl chloride, p-tosyl chloride, 4-toluenesulfonyl chloride, p-toluenesulfochloride, 4-methylbenzene-1-sulfonyl chloride, p-tolylsulfonyl chloride, p-toluenesulphonyl chloride, tosylchloride, p-toluenesulfonic acid chloride
4-methylbenzenesulfonyl chloride

Specifications

98-59-9
White
128°C
99% min
CH3C6H4SO2Cl
11,103
15,9693
Solubility in water: hydrolyses. Other solubilities: hydrolysis to acid, freely soluble in alcohol, benzene and ether
CC1=CC=C(C=C1)S(Cl)(=O)=O
190.64
190.65
Crystalline Powder
67.0°C to 69.0°C
134.0°C (10.0 mmHg)
Authentic
C7H7ClO2S
MFCD00007450
01,1179; 03,292; 04,510; 05,676; 06,598; 08,489; 11,536; 13,286
tosyl chloride, p-toluenesulfonyl chloride, p-tosyl chloride, 4-toluenesulfonyl chloride, p-toluenesulfochloride, 4-methylbenzene-1-sulfonyl chloride, p-tolylsulfonyl chloride, p-toluenesulphonyl chloride, tosylchloride, p-toluenesulfonic acid chloride
YYROPELSRYBVMQ-UHFFFAOYSA-N
4-methylbenzenesulfonyl chloride
7397
99+%
p-Toluenesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals.
Causes skin irritation.
Causes serious eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 202-684-8

Product Identifier
  • p-Toluenesulfonyl chloride

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H290-May be corrosive to metals.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only