missing translation for 'onlineSavingsMsg'
Learn More
p-Toluidine, 99%, crystalline molten mass, Thermo Scientific Chemicals Product Code.: 10128022

p-Toluidine, 99%, crystalline molten mass, Thermo Scientific Chemicals

Product Code. 10128022
1 kg, Glass bottle
Click to view available options
Quantity:
250 g
1 kg
5 kg
25 kg
Packaging:
Glass bottle
HDPE Bottle
Metal drum
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10128022

Brand: Thermo Scientific Acros 376350010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-49-0
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
InChI Key RZXMPPFPUUCRFN-UHFFFAOYSA-N
Synonym p-toluidine, 4-toluidine, p-tolylamine, 4-aminotoluene, benzenamine, 4-methyl, 4-methylbenzenamine, p-methylaniline, p-methylbenzenamine, p-toluidin, p-aminotolueneShow MoreShow Less
PubChem CID 7813
ChEBI CHEBI:37825
IUPAC Name 4-methylaniline
SMILES CC1=CC=C(C=C1)N

Specifications

Melting Point 41.0°C to 46.0°C
Color White to Brown
Boiling Point 200.0°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4NH2
Quantity 1 kg
Merck Index 15,9696
Solubility Information Solubility in water: 1.1 g/100mL. Other solubilities: freely soluble in diluted acids, freely soluble in alcohol, ether, acetone, carbon, disulfide, methanol and oils, 156% in ethanol
Formula Weight 107.16
Percent Purity 99%
Physical Form Crystalline Mass or Crystals
Assay 99%
Chemical Name or Material p-Toluidine, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0442126 Certificate TypeCertificate of Analysis Date16/11/2023 Catalog Number376350050,376350010,376350250,376352500
Lot NumberA0420948 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number376350050,376350010,376350250,376352500
Lot NumberA0404929 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number376350050,376350010,376350250,376352500
Lot NumberA0405006 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number376350050,376350010,376350250,376352500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
p-Toluidine, 99%, crystalline molten mass, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.