Learn More
Thermo Scientific™ Hypersil™ ODS C18 Columns
Use as a general purpose C18 phase for acidic and neutral analytes.
£299.08 - £3200.31
Specifications
pH 2 to 8
Pore Size 120 Å
Surface Area 170 m²/g
Temperature 60°C
USP Type L1
View More Specs
Products 54
Product Code Brand Particle Size Diameter (Metric) Length (Metric) Column Format Price Quantity & Availability  
Product Code Brand Particle Size Diameter (Metric) Length (Metric) Column Format Price Quantity & Availability  
10009432
View Documents Promotion Details
Thermo Scientific™
30105-052130
5 μm 2.1 mm 50 mm Analytical Column
£521.88
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-08-2024
to see stock.
Add to basket
11547441
View Documents Promotion Details
Thermo Scientific™
30103-034630
3 μm 4.6 mm 30 mm Analytical Column
£574.07
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
10678825
View Documents Promotion Details
Thermo Scientific™
30105-053030
5 μm 3 mm 50 mm Analytical Column
£521.88
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-08-2024
to see stock.
Add to basket
11557441
View Documents Promotion Details
Thermo Scientific™
30103-064046
3 μm 4 mm 60 mm Analytical Column
£653.35
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
16318586
View Documents Promotion Details
Thermo Scientific™
30105-259070A
5 μm 10 mm 250 mm Preparative Column, Standard
£1557.90
Each
View alternative products
Save Up to 
»
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
10288034
View Documents Promotion Details
Thermo Scientific™
30103-064630
3 μm 4.6 mm 60 mm Analytical Column
£653.35
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10728063
View Documents Promotion Details
Thermo Scientific™
30105-054630
5 μm 4.6 mm 50 mm Analytical Column
£492.97
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
16328586
View Documents Promotion Details
Thermo Scientific™
30105-259270A
5 μm 20 mm 250 mm Preparative Column, Standard
£3200.31
Each
View alternative products
Save Up to 
»
Estimated Shipment: 08-08-2024
to see stock.
Add to basket
10434593
View Documents Promotion Details
Thermo Scientific™
30105-102130
5 μm 2.1 mm 100 mm Analytical Column
£569.05
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10159014
View Documents Promotion Details
Thermo Scientific™
30105-103030
5 μm 3 mm 100 mm Analytical Column
£564.69
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
10381795
View Documents Promotion Details
Thermo Scientific™
30105-104030
5 μm 4 mm 100 mm Analytical Column
£538.94
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
10243313
View Documents Promotion Details
Thermo Scientific™
30105-104630
5 μm 4.6 mm 100 mm Analytical Column
£538.94
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10189274
View Documents Promotion Details
Thermo Scientific™
30105-122130
5 μm 2.1 mm 125 mm Analytical Column
£602.17
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-08-2024
to see stock.
Add to basket
11543181
View Documents Promotion Details
Thermo Scientific™
30105-254668
5 μm 4.6 mm 250 mm Analytical Column
£657.37
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10689965
View Documents Promotion Details
Thermo Scientific™
30105-123030
5 μm 3 mm 125 mm Analytical Column
£602.17
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
10572275
View Documents Promotion Details
Thermo Scientific™
30105-124030
5 μm 4 mm 125 mm Analytical Column
£571.05
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10783805
View Documents Promotion Details
Thermo Scientific™
30105-124630
5 μm 4.6 mm 125 mm Analytical Column
£571.05
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10263553
View Documents Promotion Details
Thermo Scientific™
30105-152130
5 μm 2.1 mm 150 mm Analytical Column
£595.14
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
10763616
View Documents Promotion Details
Thermo Scientific™
30105-153030
5 μm 3 mm 150 mm Analytical Column
£590.58
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
10429344
View Documents Promotion Details
Thermo Scientific™
30105-154030
5 μm 4 mm 150 mm Analytical Column
£571.05
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10019192
View Documents Promotion Details
Thermo Scientific™
30105-154630
5 μm 4.6 mm 150 mm Analytical Column
£566.04
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10527494
View Documents Promotion Details
Thermo Scientific™
30105-202130
5 μm 2.1 mm 200 mm Analytical Column
£629.27
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-08-2024
to see stock.
Add to basket
10498944
View Documents Promotion Details
Thermo Scientific™
30105-204030
5 μm 4 mm 200 mm Analytical Column
£595.14
Each
View alternative products
Save Up to 
»
Estimated Shipment: 05-08-2024
to see stock.
Add to basket
10456854
View Documents Promotion Details
Thermo Scientific™
30105-204630
5 μm 4.6 mm 200 mm Analytical Column
£590.58
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10140693
View Documents Promotion Details
Thermo Scientific™
30105-252130
5 μm 2.1 mm 250 mm Analytical Column
£627.46
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
10097384
View Documents Promotion Details
Thermo Scientific™
30105-253030
5 μm 3 mm 250 mm Analytical Column
£629.27
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10640584
View Documents Promotion Details
Thermo Scientific™
30105-254030
5 μm 4 mm 250 mm Analytical Column
£612.20
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10099282
View Documents Promotion Details
Thermo Scientific™
30105-254630
5 μm 4.6 mm 250 mm Analytical Column
£595.14
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10650246
View Documents Promotion Details
Thermo Scientific™
30105-013001
5 μm 3 mm 10 mm Guard Cartridge, 4 Pk.
£299.08
Pack of 4
View alternative products
Save Up to 
»
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
10581295
View Documents Promotion Details
Thermo Scientific™
30103-014001
3 μm 4 mm 10 mm Guard Cartridge, 4 Pk.
£332.20
Pack of 4
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10016024
View Documents Promotion Details
Thermo Scientific™
30103-013001
3 μm 3 mm 10 mm Guard Cartridge, 4 Pk.
£332.20
Pack of 4
View alternative products
Save Up to 
»
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
10551105
View accessory products View Documents Promotion Details
Thermo Scientific™
30103-012101
3 μm 2.1 mm 10 mm Guard Cartridge, 4 Pk.
£332.20
Pack of 4
View alternative products
Save Up to 
»
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
10414593
View accessory products View Documents Promotion Details
Thermo Scientific™
30105-014001
5 μm 4 mm 10 mm Guard Cartridge, 4 Pk.
£299.08
Pack of 4
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10543795
View accessory products View Documents Promotion Details
Thermo Scientific™
30105-012101
5 μm 2.1 mm 10 mm Guard Cartridge, 4 Pk.
£299.08
Pack of 4
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10723995
View Documents Promotion Details
Thermo Scientific™
30103-254630
3 μm 4.6 mm 250 mm Analytical Column
£657.37
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10017904
View Documents Promotion Details
Thermo Scientific™
30103-254030
3 μm 4 mm 250 mm Analytical Column
£657.37
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10649565
View Documents Promotion Details
Thermo Scientific™
30103-252130
3 μm 2.1 mm 250 mm Analytical Column
£731.51
Each
View alternative products
Save Up to 
»
Estimated Shipment: 05-08-2024
to see stock.
Add to basket
10404593
View Documents Promotion Details
Thermo Scientific™
30103-154630
3 μm 4.6 mm 150 mm Analytical Column
£627.26
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10202065
View Documents Promotion Details
Thermo Scientific™
30103-154030
3 μm 4 mm 150 mm Analytical Column
£627.26
Each
View alternative products
Save Up to 
»
Estimated Shipment: 05-08-2024
to see stock.
Add to basket
10689205
View Documents Promotion Details
Thermo Scientific™
30103-153030
3 μm 3 mm 150 mm Analytical Column
£652.33
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
10160593
View Documents Promotion Details
Thermo Scientific™
30103-152130
3 μm 2.1 mm 150 mm Analytical Column
£657.37
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
10418613
View Documents Promotion Details
Thermo Scientific™
30103-124630
3 μm 4.6 mm 125 mm Analytical Column
£627.26
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
10668334
View Documents Promotion Details
Thermo Scientific™
30103-124030
3 μm 4 mm 125 mm Analytical Column
£627.26
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10611134
View Documents Promotion Details
Thermo Scientific™
30103-104630
3 μm 4.6 mm 100 mm Analytical Column
£588.59
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10767655
View Documents Promotion Details
Thermo Scientific™
30103-104030
3 μm 4 mm 100 mm Analytical Column
£593.14
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10126383
View Documents Promotion Details
Thermo Scientific™
30103-103030
3 μm 3 mm 100 mm Analytical Column
£618.47
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
10582445
View Documents Promotion Details
Thermo Scientific™
30103-102130
3 μm 2.1 mm 100 mm Analytical Column
£623.24
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
10610944
View Documents Promotion Details
Thermo Scientific™
30103-054630
3 μm 4.6 mm 50 mm Analytical Column
£547.97
Each
View alternative products
Save Up to 
»
Estimated Shipment: 07-08-2024
to see stock.
Add to basket
10477824
View Documents Promotion Details
Thermo Scientific™
30103-053030
3 μm 3 mm 50 mm Analytical Column
£565.93
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
11553181
View Documents Promotion Details
Thermo Scientific™
30105-259070
5 μm 10 mm 250 mm Preparative Column This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11597451
View Documents Promotion Details
Thermo Scientific™
30110-154030
10 μm 4 mm 150 mm Analytical Column This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10488014
View Documents Promotion Details
Thermo Scientific™
30103-253030
3 μm 3 mm 250 mm Analytical Column This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11517451
View Documents Promotion Details
Thermo Scientific™
30105-074630
5 μm 4.6 mm 75 mm Analytical Column This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10364223
View Documents Promotion Details
Thermo Scientific™
30103-052130
3 μm 2.1 mm 50 mm Analytical Column This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Choose Thermo Scientific™ Hypersil™ ODS C18 HPLC Columns for a general purpose C18 phase to use in a broad range of applications.
Hypersil stationary phases are recognized as an industry standard in HPLC. Proven as an effective analytical tool, the classical Hypersil phases are well established and referenced in many HPLC methods worldwide. Quality is assured by manufacture under ISO 9001-2000 quality guidelines.
For more challenging separations that require improved chromatographic performance, we recommend the use of Thermo Scientific™ Hypersil GOLD™ columns as an alternative.
  • High efficiency and proven reproducibility
  • Exceptionally reliable
  • Separation of a wide range of compounds including nonpolar, moderately polar and lipophilic compounds like triglycerides
Specifications
Specifications
2 to 8
170 m²/g
L1
10%
Yes
120 Å
60°C
C18
Reversed Phase
Reversed Phase
Videos
SDS
Documents
Documents
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.