missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Centrifuge Tube Mini-Cooler
Designed to protect a wide range of solutions (enzymes, DNA, RNA, cell suspensions) by helping to maintain freezer temperatures on the lab bench.
£143.45 - £188.00
Specifications
For Use With (Equipment) | Centrifuge tube |
---|---|
Material | Polycarbonate |
Type | Mini Cooler |
Capacity (Metric) | 0.5 to 2 mL |
Holds | 0.5 to 2 mL Tubes |
Product Code | Brand | Color | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Description | Price | Quantity & Availability | ||||
10787281
|
BRAND™
114935 |
Yellow | Maintains −20°C temperature for 60 min. |
£162.02
Each |
In Stock
Log in to see stock. |
||||
10141921
|
BRAND™
114930 |
Red | Maintains 0°C temperature for 60 min. |
£188.00
Each |
In Stock
Log in to see stock. |
||||
10441063
|
BRAND™
114940 |
White | Maintains −70°C temperature for 45 min. |
£143.45
Each |
In Stock
Log in to see stock. |
||||
Description
- Manufactured from durable polycarbonate
- Filled with non-toxic gel
- Hold twelve 0.5 to 2.0mL tubes
- Provided individually
Specifications
Centrifuge tube | |
Mini Cooler | |
0.5 to 2 mL Tubes |
Polycarbonate | |
0.5 to 2 mL |