Learn More
Thermo Scientific™ Heratherm™ Refrigerated Incubators
Offers precise temperature control, increased energy efficiency, and ease of usability. Equipped with Peltier technology to reach and maintain the precise ambient temperatures you need for reliable results.
£6834.85 - £10417.92
Specifications
Chamber Material | Stainless Steel (304/1.4301) |
---|---|
Material | Coated Steel |
Temperature Range (Metric) | 5°C to 70°C |
Type | Automated Incubator |
Display | Large Vacuum Fluorescent |
Product Code | Brand | Capacity (English) | Capacity (Metric) | Voltage | Includes | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (English) | Capacity (Metric) | Voltage | Includes | Price | Quantity & Availability | ||
15689408
|
Thermo Scientific™
51031564 |
6.3 cu. ft. | 178 L | 230 V | Internal Electrical Outlet: EU (CEE 7/4 plug and interior outlet) |
£7576.59
Each |
Estimated Shipment: 04-09-2024
Log in to see stock. |
||
15608540
|
Thermo Scientific™
51031562 |
6.3 cu. ft. | 178 L | 100/230 V | Base Model |
£6834.85
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||
15662079
|
Thermo Scientific™
51031567 |
13.5 cu. ft. | 381 L | 230 V | Internal Electrical Outlet: EU (CEE 7/4 plug and interior outlet) |
£10417.92
Each |
Estimated Shipment: 19-08-2024
Log in to see stock. |
||
Description
Achieve untapped potential in incubation with the temperature precision you need for applications that demand it. With a temperature range of +5 to +70°C, Thermo Scientific™ Heratherm™ Refrigerated Incubators are equipped with Peltier technology to reach and maintain the precise ambient temperatures you need for reliable results. With no hazardous refrigerants, these units use less energy than the conventional product, making it the ideal choice for energy-conscious labs.
- Temperature range +5 to +70°C
- Choose benchtop model 178L / 6.3 cu.ft. or floor model with casters 381L / 13.5 cu. ft.
- Great temperature uniformity and stability to keep the samples in a safe environment - uniformity as low as +/-0.3°C (20°C), stability as low as +/-0.1°C
- Energy consumption as low as 80Wh/h (benchtop) / 150Wh/h (floor) at 20°C - Note that comparable compressor cooled units consume up to 6 times this amount*
- CFC free / HFC free polyurethane insulation foam for outstanding temperature performance and condensation prevention
- Heated door with intelligent energy control to ensure no condensation – even at low temperatures
- Sophisticated timer function with weekly, daily or real time mode
- Intuitive programmability function for defined temperature ramping, soak/cool and dwell: store up to 10 programs with 10 easy steps
- Adjustable over temperature shut-off for protocols that require setting of over temperature for protection – (class 2 according to norm DIN12880)
- Safe viewing of samples through internal glass door with limited impact on temperature
- Safe containment with automatic over and under temperature alarm
- Standard access port for data monitoring (large diameter of 42mm / 1.65 in.)
- Flexible shelf system for optimal loading, easy to remove for cleaning, with non-tip protection
- Stainless steel interior (1.4301 / ASTM 304) and rounded corners for easy cleaning
Warranty
2 years
Specifications
Stainless Steel (304/1.4301) | |
5°C to 70°C | |
Large Vacuum Fluorescent | |
Energy efficient | |
1 | |
Peltier technology: no refrigerant |
Coated Steel | |
Automated Incubator | |
Solid outer door & internal glass door | |
CFC free/HFC free polyurethane insulation foam | |
1.6 in. (42mm) |