Learn More
Sartorius™ Entris™ II Essential Precision Balance
Entris™ II Essential line of balances offers highly accurate results via monolithic weigh cell technology, fastest stabilization time in its class using state-of -the-art weighing sensors and high chemical resistance.
£945.00 - £1910.00
Specifications
Certifications/Compliance | CSA number 80000336/CE certificate |
---|---|
Connection Type | 9 Pin USB C |
Display Type | LED; Intuitive, Wear Resistant with Touch Technology |
Includes | Pan support, Frame draft shield, Power supply unit with country-specific AC adapters, Operating Instructions, Shock absorber |
Material | PBT, Glass, Stainless Steel |
Product Code | Brand | Capacity | Readability | Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Readability | Type | Price | Quantity & Availability | |||
16344698
|
Sartorius™
BCE223-1S |
220 g | 1 mg | No Internal Calibration |
£1055.00
1 unit |
Estimated Shipment: 20-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16374698
|
Sartorius™
BCE223I-1S |
220 g | 1 mg | Internal Calibration |
£1275.00
1 unit |
Estimated Shipment: 20-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16364708
|
Sartorius™
BCE323-1S |
320 g | 1 mg | No Internal Calibration |
£1160.00
1 unit |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16394708
|
Sartorius™
BCE323I-1S |
320 g | 1 mg | Internal Calibration |
£1390.00
1 unit |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16344718
|
Sartorius™
BCE423-1S |
420 g | 1 mg | No Internal Calibration |
£1280.00
1 unit |
Estimated Shipment: 14-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16374718
|
Sartorius™
BCE423I-1S |
420 g | 1 mg | Internal Calibration |
£1490.00
1 unit |
Estimated Shipment: 20-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16324748
|
Sartorius™
BCE653-1S |
600 g | 1 mg | No Internal Calibration |
£1775.04
1 unit |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16354748
|
Sartorius™
BCE653I-1S |
600 g | 1 mg | Internal Calibration |
£1910.00
1 unit |
Estimated Shipment: 20-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16344738
|
Sartorius™
BCE623-1S |
620 g | 1 mg | No Internal Calibration |
£1500.00
1 unit |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16374738
|
Sartorius™
BCE623I-1S |
620 g | 1 mg | Internal Calibration |
£1690.00
1 unit |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16304738
|
Sartorius™
BCE622-1S |
620 g | 10 mg | No Internal Calibration |
£945.00
1 unit |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16334738
|
Sartorius™
BCE622I-1S |
620 g | 10 mg | Internal Calibration |
£1150.00
1 unit |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16344758
|
Sartorius™
BCE822-1S |
820 g | 10 mg | No Internal Calibration |
£1050.00
1 unit |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16374758
|
Sartorius™
BCE822I-1S |
820 g | 10 mg | Internal Calibration |
£1270.00
1 unit |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16384678
|
Sartorius™
BCE1202-1S |
1200 g | 10 mg | No Internal Calibration |
£1165.00
1 unit |
Estimated Shipment: 20-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16314688
|
Sartorius™
BCE1202I-1S |
1200 g | 10 mg | Internal Calibration |
£1395.00
1 unit |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16304698
|
Sartorius™
BCE2202-1S |
2200 g | 10 mg | No Internal Calibration |
£1285.00
1 unit |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16334698
|
Sartorius™
BCE2202I-1S |
2200 g | 10 mg | Internal Calibration |
£1495.00
1 unit |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16364688
|
Sartorius™
BCE2201-1S |
2200 g | 100 mg | No Internal Calibration |
£1050.00
1 unit |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16394688
|
Sartorius™
BCE2201I-1S |
2200 g | 100 mg | Internal Calibration |
£1270.00
1 unit |
Estimated Shipment: 20-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16324708
|
Sartorius™
BCE3202-1S |
3200 g | 10 mg | No Internal Calibration |
£1400.00
1 unit |
Estimated Shipment: 28-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16354708
|
Sartorius™
BCE3202I-1S |
3200 g | 10 mg | Internal Calibration |
£1595.00
1 unit |
Estimated Shipment: 14-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16304718
|
Sartorius™
BCE4202-1S |
4200 g | 10 mg | No Internal Calibration |
£1505.00
1 unit |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16334718
|
Sartorius™
BCE4202I-1S |
4200 g | 10 mg | Internal Calibration |
£1695.00
1 unit |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16384718
|
Sartorius™
BCE5201-1S |
5200 g | 100 mg | No Internal Calibration |
£1160.00
1 unit |
Estimated Shipment: 20-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16314728
|
Sartorius™
BCE5201I-1S |
5200 g | 100 mg | Internal Calibration |
£1390.00
1 unit |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16364728
|
Sartorius™
BCE6202-1S |
6200 g | 10 mg | No Internal Calibration |
£1775.04
1 unit |
Estimated Shipment: 15-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16394728
|
Sartorius™
BCE6202I-1S |
6200 g | 10 mg | Internal Calibration |
£1910.00
1 unit |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16324728
|
Sartorius™
BCE6200-1S |
6200 g | 1000 mg | No Internal Calibration |
£1050.00
1 unit |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16354728
|
Sartorius™
BCE6200I-1S |
6200 g | 1000 mg | Internal Calibration |
£1270.00
1 unit |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16304758
|
Sartorius™
BCE8201-1S |
8200 g | 100 mg | No Internal Calibration |
£1285.00
1 unit |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16334758
|
Sartorius™
BCE8201I-1S |
8200 g | 100 mg | Internal Calibration |
£1490.00
1 unit |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16364748
|
Sartorius™
BCE8200-1S |
8200 g | 1000 mg | No Internal Calibration |
£1160.00
1 unit |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16394748
|
Sartorius™
BCE8200I-1S |
8200 g | 1000 mg | Internal Calibration |
£1390.00
1 unit |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Entris™ II Essential line of balances offers highly accurate results via monolithic weigh cell technology, fastest stabilization time in its class using state-of -the-art weighing sensors and high chemical resistance.
- High quality standards engineered in Germany
- Excellent repeatability with rectangular weighing pan
- Overload protection
- Rugged design weighs up to a pre-set amount
- High chemical resistance
- Crafted from hard-wearing polybutylene terephthalate (PBT), stainless steel and glass
- Prevents cross contamination
- Wipe-clean design
- Easy-to-remove parts
- Effective draft shield
- Minimizes weighing errors caused by electrostatically charged samples using specially coated glass parts
- Some models are equipped with isoCAL (internal calibration and adjustment) for total assurance of accurate weighing results
- Assures (SOP) compliant operation with self-notification if calibration is outside the normal range
- Combines intuitive, wear resistant LED and touch technology
- Plug-and-play convenience
- Data output for dynamic weighing applications
- Easily adapts to your ambient conditions
- 12 built-in applications
- GLP / GMP compliant printout
- Modern connectivity methods
- Password protection for secure operation
Specifications
CSA number 80000336/CE certificate | |
LED; Intuitive, Wear Resistant with Touch Technology | |
PBT, Glass, Stainless Steel | |
219 mm |
9 Pin USB C | |
Pan support, Frame draft shield, Power supply unit with country-specific AC adapters, Operating Instructions, Shock absorber | |
Electronic Toploading Balances | |
100 to 240 VAC (±10%) |