Learn More
Cytiva (formerly Pall Lab) Macrosep™ Advance Centrifugal Devices with Omega Membrane
Macrosep Advance Centrifugal Devices with Omega Membrane quickly concentrates up to 20 mL of biological sample.
£241.00 - £1105.00
Specifications
Description | EFA: 7.2 cm2 |
---|---|
Material (Membrane) | PES |
Sterility | Non-sterile |
Volume (Metric) | 5 to 20 mL (Sample) |
Product Code | Brand | MWCO | Includes | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Includes | Color | Price | Quantity & Availability | |||
17154731
|
Cytiva (formerly Pall Lab)
MAP003C38 |
3 kDa | 100/Pk. | Gray |
£985.00
Pack of 100 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
|||
17174731
|
Cytiva (formerly Pall Lab)
MAP010C38 |
10 kDa | 100/Pk. | Blue |
£985.00
Pack of 100 |
Estimated Shipment: 03-10-2024
Log in to see stock. |
|||
17124731
|
Cytiva (formerly Pall Lab)
MAP001C37 |
1 kDa | 24/Pk. | Yellow |
£241.00
Pack of 24 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
|||
17184731
|
Cytiva (formerly Pall Lab)
MAP030C37 |
30 kDa | 24/Pk. | Red |
£242.00
Pack of 24 |
Estimated Shipment: 03-10-2024
Log in to see stock. |
|||
17104741
|
Cytiva (formerly Pall Lab)
MAP100C37 |
100 kDa | 24/Pk. | Clear |
£306.00
Pack of 24 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||
17134731
|
Cytiva (formerly Pall Lab)
MAP001C38 |
1 kDa | 100/Pk. | Yellow |
£1105.00
Pack of 100 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
|||
17164731
|
Cytiva (formerly Pall Lab)
MAP010C37 |
10 kDa | 24/Pk. | Blue |
£293.00
Pack of 24 |
In Stock
Log in to see stock. |
|||
17114741
|
Cytiva (formerly Pall Lab)
MAP100C38 |
100 kDa | 100/Pk. | Clear |
£1105.00
Pack of 100 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
|||
17194731
|
Cytiva (formerly Pall Lab)
MAP030C38 |
30 kDa | 100/Pk. | Red |
£985.00
Pack of 100 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
|||
17144731
|
Cytiva (formerly Pall Lab)
MAP003C37 |
3 kDa | 24/Pk. | Gray |
£425.00
Pack of 24 |
Estimated Shipment: 03-10-2024
Log in to see stock. |
|||
Description
- Exchange buffer or remove salt of chromatography eluates and gradient fractions
- For research applications only
Specifications
EFA: 7.2 cm2 | |
PES | |
Non-sterile | |
5 to 20 mL (Sample) |