Learn More
Fisherbrand™ Amber Borosilicate Glass Class A Volumetric Flask with Stopper
Amber Borosilicate Glass Volumetric Flask includes interchangeable polyethylene stopper.
£52.35 - £377.71
Product Code | Brand | Closure Size | Capacity (Metric) | Tolerance | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Capacity (Metric) | Tolerance | Price | Quantity & Availability | |||
11359443
|
Fisherbrand™
|
24/29 | 1000 mL | ±0.4 mL |
£316.00
Pack of 2 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
11379433
|
Fisherbrand™
|
10/19 | 10 mL | ±0.04 mL |
£377.71
Pack of 5 |
In Stock
Log in to see stock. |
|||
11329443
|
Fisherbrand™
|
14/23 | 200 mL | ±0.15 mL |
£174.41
Pack of 2 |
In Stock
Log in to see stock. |
|||
11339443
|
Fisherbrand™
|
14/23 | 250 mL | ±0.15 mL |
£173.38
Pack of 2 |
In Stock
Log in to see stock. |
|||
11389433
|
Fisherbrand™
|
10/19 | 20 mL | ±0.04 mL |
£206.00
Pack of 5 |
In Stock
Log in to see stock. |
|||
11399433
|
Fisherbrand™
|
10/19 | 25 mL | ±0.04 mL |
£203.30
Pack of 5 |
In Stock
Log in to see stock. |
|||
11349443
|
Fisherbrand™
|
19/24 | 500 mL | ±0.25 mL |
£213.00
Pack of 2 |
In Stock
Log in to see stock. |
|||
11309443
|
Fisherbrand™
|
12/21 | 50 mL | ±0.06 mL |
£251.81
Pack of 5 |
In Stock
Log in to see stock. |
|||
11319443
|
Fisherbrand™
|
12/21 | 100 mL | ±0.01 mL |
£243.55
Pack of 5 |
In Stock
Log in to see stock. |
|||
10682842
|
Fisherbrand™
|
12/21 | 50 mL | ±0.06 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Amber Borosilicate Glass Volumetric Flask includes interchangeable polyethylene stopper.
- With one mark graduation and interchangeable polyethylene stopper.
- The details on each flask include confirmation of compliance to International Standards as well as an individual re-order code.
- The Fisherbrand 'Batch Certificate of Compliance' is downloadable for this item from the 'Documentation' tab
Specifications
Amber | |
Borosilicate Glass | |
Yes | |
A |