Learn More
Thermo Scientific™ PageRuler™ Prestained Protein Ladder, 10 to 180 kDa
Thermo Scientific PageRuler Prestained Protein Ladder is a mixture of 10 blue-, orange-, and green-stained proteins (10 to 180 kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and western blotting.
Brand: Thermo Scientific™ 26616
115.52 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Thermo Scientific PageRuler Prestained Protein Ladder is a mixture of 10 blue-, orange-, and green-stained proteins (10 to 180 kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and western blotting. The protein ladder is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.
Applications
- Monitoring protein migration during SDS-polyacrylamide gel electrophoresis
- Monitoring protein transfer onto membranes after western blotting
- Sizing of proteins on SDS-polyacrylamide gels and western blots

Specifications
Contents: two vials of 250 μL each |
|
10 | |
Yes | |
3 colors: Blue, Orange, Green | |
180, 130, 100, 70, 55, 40, 35, 25, 15, 10 kDa | |
2 x 250 μL | |
Western Blotting, SDS-PAGE |
Colorimetric, NIR Fluorescence (700 nm), RGB Fluorescence (555 nm) | |
Protein Ladder | |
10 to 180 kDa | |
Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels | |
PageRuler™ | |
Approved for shipment on Wet or Dry Ice |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2800371 | Certificate of Analysis | 29/01/2025 | ,26616,26616_LITL18 PROMO |
2812403 | Certificate of Analysis | 29/01/2025 | ,26616,26616_LITL18 PROMO |
2827769 | Certificate of Analysis | 29/01/2025 | ,26616,26616_LITL18 PROMO |
2828767 | Certificate of Analysis | 29/01/2025 | ,26616,26616_LITL18 PROMO |
2809291 | Certificate of Analysis | 29/01/2025 | ,26616,26616_LITL18 PROMO |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.