missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Papain, Carica Papaya Latex

A cysteine protease; breaks down intercellular matrix of cartilage

£61.31 - £984.90

Chemical Identifiers

CAS 9001-73-4
Synonym EC 3.4.22.2
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15404369
View Documents Promotion Details
Thermo Scientific Chemicals
J61875.MB
25 mg
£61.31
25mg
In Stock
to see stock.
Add to basket
 
15414369
View Documents Promotion Details
Thermo Scientific Chemicals
J61875.MC
100 mg
£131.66
100mg
In Stock
to see stock.
Add to basket
 
15494359
View Documents Promotion Details
Thermo Scientific Chemicals
J61875.03
1 g
£984.90
1g
Estimated Shipment: 12-12-2024
to see stock.
Add to basket
 
Description

Description

Papain, from papaya latex, has been used to digest carotid artery smooth muscle cells. Papain is used in dissecting solutions. It is used to produce Fab fragments of antibodies. It is used for cell dissociation since it has been shown to be more effective and less damaging with certain tissues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9001-73-4
EC 3.4.22.2

Specifications

White to Off-white
23 kd
2°C to 8°C
EC 3.4.22.2
A cysteine protease; breaks down intercellular matrix of cartilage
9001-73-4
Lyophilized powder,23 kDa
Protease
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 232-627-2

Recommended Storage : Keep cold

Product Identifier
  • Papain, Carica Papaya Latex

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Papain

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only