Learn More
Thermo Scientific™ PathoDx Strep Grouping Universal Kit
Support the functionality of the Thermo Scientific™ PathoDx™ Strep Grouping Kit with Thermo Scientific™ PathoDx™ Strep Grouping Universal Kit.
Brand: Thermo Scientific™ R62076
131.94 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Support the functionality of the Thermo Scientific™ PathoDx™ Strep Grouping Kit with Thermo Scientific™ PathoDx™ Strep Grouping Universal Kit. This kit is a set of components to be used with selected, separately purchased PathoDxStrep Grouping Reagents designed to identify beta-haemolytic streptococci of Lancefield Groups A, B, C, F or G from primary culture plates.This accessory kit contains: Extraction reagent 1; Extraction reagent 2; Extraction reagent 3; Positive control; Reaction cards; Stirrers.
Includes:
Control antigens ABC and FG; extraction reagents 1, 2, 3; stirrers; disposable slides
Requires:
For use with Part No. R62030, R62031, R62032, R62034, R62035.

Specifications
Positive | |
beta-hemolytic streptococci (Groups A, B, C, F, G) | |
Control antigens ABC and FG; extraction reagents 1, 2, 3; stirrers; disposable slides | |
60 Tests/Kit |
PathoDX™ Strep Universal Grouping Kit | |
Streptococcus Test Kit | |
Lancefield A, B, C, D, F and G |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- PathoDX STREP Grouping Universal Kit
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 1 A
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Sodium nitrite, Acetic acid, Carbonic acid
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.