Learn More
Gibco™ PBS, pH 7.4

Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.
Brand: Gibco™ 10010056
86.67 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.
117.36 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation.
We offer a variety of Gibco™ PBS formulations for a range of cell culture applications.
This PBS is manufactured as follows:
Without
• Calcium
• Magnesium
• Phenol Red
The complete formulation is available.

Specifications
15°C to 30°C | |
Phosphate Buffered Saline (PBS) | |
Mammalian Cell Culture | |
7.3 to 7.5 | |
CE-IVD | |
Sterile-filtered | |
cGMP-compliant under the ISO 13485 standard |
1 X | |
10 x 500 mL | |
280 - 315 mOsm/kg | |
No Phenol Red | |
Ambient | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.