missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PBS, pH 7.4
GREENER_CHOICE

Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.

£14.30 - £450.00

Specifications

Content And Storage 15°C to 30°C
Dilution 1 X
Product Type Phosphate Buffered Saline (PBS)
For Use With (Application) Mammalian Cell Culture
Osmolality 280 - 315 mOsm/kg
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11593377
View Documents
Gibco™
10010015
500 mL
£14.30
500mL
Please to purchase this item. Need a web account? Register with us today!
11503387
GREENER_CHOICE
View Documents
Gibco™
10010056
10 x 500 mL
£124.00
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
15390314
View Documents
Gibco™
10010002
5 L
£171.00
5 L
Please to purchase this item. Need a web account? Register with us today!
15316409
View Documents
Gibco™
10010001
10 L
£450.00
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation.

We offer a variety of Gibco™ PBS formulations for a range of cell culture applications.

This PBS is manufactured as follows:

Without
• Calcium
• Magnesium
• Phenol Red

The complete formulation is available.

Specifications

Specifications

15°C to 30°C
Phosphate Buffered Saline (PBS)
280 - 315 mOsm/kg
No Phenol Red
Sterile-filtered
cGMP-compliant under the ISO 13485 standard
1 X
Mammalian Cell Culture
7.3 to 7.5
Ambient
Liquid
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 36 Months

SDS
Documents

Documents

Product Certifications

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.