Learn More
Thermo Scientific™ Urea
Pure, crystalline urea (CH4N2O), CAS 57-13-6; denaturant with low UV-absorptivity for protein biology methods (elution, solubilization, refolding, storage).
Brand: Thermo Scientific™ 29700
49.64 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Thermo Scientific™ Pierce Urea is pure, crystalline urea, a protein denaturant with low UV-absorptivity.
Additional Information and Online Ordering
Highlights:
- Melting point: 132-136°C
- Specification: A280 <0.100

Specifications
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C. | |
Chaotrope, Denaturant | |
Urea |
Powder | |
1 kg |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.