Learn More
Thermo Scientific™ 8M Guanidine-HCl Solution
Ready-to-use, 8M solution of guanidinium hydrochloride (GdnCl, GuHCl), CAS 50-01-1; dilute and pH-adjust to any concentration for protein biology methods.
Brand: Thermo Scientific™ 24115
326.80 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Thermo Scientific™ Pierce 8M Guanidine-HCl Solution is an accurately formulated, ready-to-use solution of guanidine hydrochloride, that can be easily diluted and pH-adjusted to any concentration below 8M.
Highlights:
- Any concentration below 8 M can be quickly and easily prepared
- No more wasting valuable research time handling difficult-to-weigh crystals for slow-to-prepare guanidine·HCl solutions
- Free of UV-absorbing materials at 225-300nm
- Sharp UV cut-off spectrum with OD260 < 0.03
- Typical metals: Cu < 0.02ppm, Zn < 0.15ppm, Fe < 0.25ppm, Pb < 0.005ppm
- Particulate-free, crystal-clear solution
- Aldehyde-free
- Excellent stability

Specifications
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area. | |
Chaotrope, Denaturant | |
Guanidine Hydrochloride |
Liquid | |
200 mL |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- 8 M Guanidine
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P270-Do not eat, drink or smoke when using this product.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : guanidinium chloride
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.