missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva PEEK Sample Loops
Resists organic solvents and salt buffers commonly used in chromatography of biomolecules. Cytiva Life Sciences™ PEEK Sample Loops are an important accessory for chromatography applications.
Brand: Cytiva 18112039
This item is not returnable.
View return policy
Description
- Wetted parts are resistant to organic solvents
- Wetted parts are resistant to salt buffers
- Except 100 % Ethyl acetate, 100 % Hexane and 100 % Tetrahydrofuran
- Polyetheretherketone (PEEK)

Specifications
Sample Loop | |
1.58 mm | |
10 μL |
PEEK | |
0.25 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva PEEK Sample Loops > Inner Diameter: 0.25mm; Volume: 10μL
Spot an opportunity for improvement?Share a Content Correction