Learn More
Thermo Scientific™ NsbI (FspI) (10 U/μL)
Cut at TGC^GCA sites with NsbI (FspI) restriction enzyme, which performs best at 37°C in Tango buffer (Isoschizomers: Acc16I, AviII, FspI).
Brand: Thermo Scientific™ ER1221
22.33 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Lambda DNA digested with NsbI (FspI), 0.7% agarose, 15 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
Highlights
- Superior quality -stringent quality control and industry leading manufacturing process
- Convenient color-coded Five Buffer System
- Includes universal Tango buffer for double-digestions
- BSA premixed in reaction buffers
- Wide selection of restriction endonuclease specificities

Specifications
10 U/μL | |
NsbI (FspI) | |
Yes | |
No | |
Restriction Enzyme |
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive | |
10X Buffer Tango | |
37°C | |
400 units | |
Traditional Cloning |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.