missing translation for 'onlineSavingsMsg'
Learn More
Pentafluorophenyl 6-(tetrahydropyran-4-yloxy)nicotinate, 97%, Thermo Scientific™ Product Code.: 11356815

Pentafluorophenyl 6-(tetrahydropyran-4-yloxy)nicotinate, 97%, Thermo Scientific™

Product Code. 11356815
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 11356815

Brand: Thermo Scientific Maybridge CC58326CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 910036-96-3
Molecular Formula C17H12F5NO4
Molecular Weight (g/mol) 389.28
MDL Number MFCD09702389
InChI Key DOADFHHFWSVLLM-UHFFFAOYSA-N
Synonym pentafluorophenyl 6-tetrahydropyran-4-yloxy nicotinate, 2,3,4,5,6-pentafluorophenyl 6-oxan-4-yloxy pyridine-3-carboxylate, pentafluorophenyl 6-oxan-4-yloxy pyridine-3-carboxylate, pentafluorophenyl 6-oxan-4-yl oxy pyridine-3-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl 6-oxan-4-yloxy pyridine-3-carboxylate, 6-4-oxanyloxy-3-pyridinecarboxylic acid 2,3,4,5,6-pentafluorophenyl esterShow MoreShow Less
PubChem CID 24229582
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 6-(oxan-4-yloxy)pyridine-3-carboxylate
SMILES FC1=C(F)C(F)=C(OC(=O)C2=CN=C(OC3CCOCC3)C=C2)C(F)=C1F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 389.27
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 6-(tetrahydropyran-4-yloxy)nicotinate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pentafluorophenyl 6-(tetrahydropyran-4-yloxy)nicotinate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.